Order Entry
Export
Orders LinkContactUsLinkComponent
3,5-Dinitrobenzonitrile ≥98%

3,5-Dinitrobenzonitrile ≥98%

Glass bottle for solids, 25 g
Catalog # A15223.14
Toxic
Health
Irritant
3,5-Dinitrobenzonitrile ≥98%
Catalog # A15223.14
CAS Number:  4110-35-4
3,5-Dinitrobenzonitrile ≥98%
3,5-Dinitrobenzonitrile ≥98%

Glass bottle for solids, 25 g

CAS Number:  4110-35-4
Price
128,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 4110-35-4 Molecular Formula: C7H3N3O4 Formula Weight: 193.12 Melting Point: 127-130° UN No.: 3439 MDL No.: MFCD00007228

  • CAS Number:4110-35-4
  • Formula:C₇H₃N₃O₄
  • MDL Number:MFCD00007228
  • Molecular Weight:193.12 g/mol
  • Melting Pt:127…130 °C
  • Storage Temperature:Ambient
  • EINECS:223-889-9
  • UN:3439
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications