Orders LinkContactUsLinkComponent
3,5-Dinitrobenzonitrile ≥98%
Glass bottle for solids, 5 g
  A15223.06
3,5-Dinitrobenzonitrile ≥98%
  A15223.06
 :  4110-35-4
3,5-Dinitrobenzonitrile ≥98%
3,5-Dinitrobenzonitrile ≥98%
Glass bottle for solids, 5 g
 :  4110-35-4
42,60 €
1 * 5 g
Purity: 98% CAS No.: 4110-35-4 Molecular Formula: C7H3N3O4 Formula Weight: 193.12 Melting Point: 127-130° UN No.: 3439 MDL No.: MFCD00007228
  •  :4110-35-4
  •  :C₇H₃N₃O₄
  •  :MFCD00007228
  •  :193.12 g/mol
  •  :127…130 °C
  •  :Ambient
  •  :223-889-9
  •  :3439
  •  :6.1,III
  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g