Trypan blue 60% (dye content)
25 g
Supplier: Thermo Fisher Scientific
Trypan blue 60% (dye content)
Catalog # A18600.14
Supplier: Thermo Fisher Scientific
CAS Number: 72-57-1
Jump to:
Product Details & Documents
CAS No.: 72-57-1 Molecular Formula: C34H24N6Na4O14S4 Formula Weight: 960.82 Melting Point: 300° Merck Reference: 14,9792 MDL No.: MFCD00003969
Used in dye exclusion studies in cell culture
Synonyms: Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt, CI 23850, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt, Benzamine blue, Trypan blue sodium salt, Direct Blue 14, Diphenyl Blue, AzidinBlue, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulphonate)
- CAS Number:72-57-1
- Formula:C₃₄H₂₄N₆Na₄O₁₄S₄
- MDL Number:MFCD00003969
- Molecular Weight:960.82 g/mol
- Merck Index:14,9792
- Melting Pt:>300 °C
- Storage Temperature:Ambient
- EINECS:200-786-7
Specifications
Specifications
- Pk:25 g
- Cat. No.:A18600.14



