Order Entry
Export
Orders LinkContactUsLinkComponent
Thread adapters for pumps

Thread adapters for pumps

Extrenal thread adapter, PP, black, 61 mm - 50,8 mm BSP
Catalog # BURK0600-3007
Supplier:   BURKLE
Thread adapters for pumps
Catalog # BURK0600-3007
Supplier:  BURKLE
Thread adapters for pumps
Thread adapters for pumps

Extrenal thread adapter, PP, black, 61 mm - 50,8 mm BSP

Price
12,40 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    Extrenal thread adapter, PP, black, 61 mm - 50,8 mm BSP