Order Entry
Export
Orders LinkContactUsLinkComponent
n-Dodecyl-β-D-maltoside

n-Dodecyl-β-D-maltoside

Glass bottle for solids, 5 g
Catalog # PIER89903
Irritant
n-Dodecyl-β-D-maltoside
Catalog # PIER89903
CAS Number:  69227-93-6
n-Dodecyl-β-D-maltoside
n-Dodecyl-β-D-maltoside

Glass bottle for solids, 5 g

CAS Number:  69227-93-6
Price
638,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  DDM, Lauryl-β-D-maltoside, Dodecyl-β-D-maltopyranoside, (2R,3R,4S,5S,6R)-2-(((2R,3S,4R,5R,6R)-6-(Dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, N-Dodecyl-Beta-Maltoside
  • CAS Number:69227-93-6
  • Formula:C₂₄H₄₆O₁₁
  • MDL Number:MFCD00043012
  • Molecular Weight:510.6 g/mol
  • Storage Temperature:Freezer
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications