Wash buffer, Pierce™
Pierce™ borate buffer, 20X, 8.5, 0,5 L
PIER28341
These wash buffers are designed for use in dialysis, cross-linking, enzyme assays, ELISAs, immunohistochemistry, protein plate-coating, biotinylation applications.
- Ready to use without having to reconstitute with ultrapure water
- Eliminates the possibility of powder remaining in a packet
- 10X or 20X concentration eliminates time spent waiting for powder to dissolve
- Storage as concentrated stock minimizes bench space needed for large volume solutions
- Pk:0,5 L
- Description:Pierce™ borate buffer, 20X
- pH value:8.5



