L-Glutamine, HyClone™
L-Glutamine 200 mM solution, concentration 29,2 mg/ml, 100 ml
Supplier: HyClone products (Cytiva)
Jump to:
Product Details & Documents
HyClone™ L-glutamine is used as a supplement in serum- and protein-free media. It supports the growth of cells that have high energy demands and synthesise large amounts of proteins and nucleic acids.
Documents
Specifications
Specifications
- Pk:100 ml
- Description:L-Glutamine 200 mM solution, concentration 29,2 mg/ml