Order Entry
Export
Orders LinkContactUsLinkComponent
5-Bromo-2-chlorophenol ≥98%

5-Bromo-2-chlorophenol ≥98%

Glass bottle for solids, 1 g
Catalog # B25452.03
Irritant
5-Bromo-2-chlorophenol ≥98%
Catalog # B25452.03
CAS Number:  183802-98-4
5-Bromo-2-chlorophenol ≥98%
5-Bromo-2-chlorophenol ≥98%

Glass bottle for solids, 1 g

CAS Number:  183802-98-4
Price
62,80 €
Log in for availability
1 * 1 g
Product Details & Documents

Purity: 98+% CAS No.: 183802-98-4 Molecular Formula: C6H4BrClO Formula Weight: 207.46 Melting Point: 56-58° MDL No.: MFCD00672940

  • CAS Number:183802-98-4
  • Formula:C₆H₄BrClO
  • MDL Number:MFCD00672940
  • Molecular Weight:207.45 g/mol
  • Melting Pt:56…58 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications