Order Entry
Export
Orders LinkContactUsLinkComponent
2-Hydroxycyclohexanone dimer ≥90%

2-Hydroxycyclohexanone dimer ≥90%

Glass bottle for solids, 25 g
Catalog # H27199.14
Flammable
Irritant
2-Hydroxycyclohexanone dimer ≥90%
Catalog # H27199.14
CAS Number:  30282-14-5
2-Hydroxycyclohexanone dimer ≥90%
2-Hydroxycyclohexanone dimer ≥90%

Glass bottle for solids, 25 g

CAS Number:  30282-14-5
Price
200,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 90+% CAS No.: 30282-14-5 Molecular Formula: C12H20O4 Formula Weight: 228.28 Flash Point: 79°(174°F) Melting Point: 100-107° Boiling Point: 83-86°/13mm UN No.: 1325 MDL No.: MFCD00151157

  • CAS Number:30282-14-5
  • Formula:C₆H₁₀O₂
  • MDL Number:MFCD00151157
  • Molecular Weight:228.28 g/mol
  • Boiling Pt:83…86 °C (13 mmHg)
  • Melting Pt:100…107 °C
  • Flash Pt:79 °C (174 °F)
  • Storage Temperature:Ambient
  • UN:1325
  • ADR:4.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications