Order Entry
Export
Orders LinkContactUsLinkComponent
2-Hydroxycarbazole ≥97%

2-Hydroxycarbazole ≥97%

Glass bottle for solids, 25 g
Catalog # H27765.14
Irritant
2-Hydroxycarbazole ≥97%
Catalog # H27765.14
CAS Number:  86-79-3
2-Hydroxycarbazole ≥97%
2-Hydroxycarbazole ≥97%

Glass bottle for solids, 25 g

CAS Number:  86-79-3
Price
732,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 97% CAS No.: 86-79-3 Molecular Formula: C12H9NO Formula Weight: 183.21 Odour: Odourless Melting Point: 262-273° MDL No.: MFCD00004962

Synonyms:  Carbazol-2-ol
  • CAS Number:86-79-3
  • Formula:C₁₂H₉NO
  • MDL Number:MFCD00004962
  • Molecular Weight:183.21 g/mol
  • Melting Pt:262…273 °C
  • Storage Temperature:Ambient
  • EINECS:201-699-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications