Order Entry
Export
Orders LinkContactUsLinkComponent
Lead(II) sulphide (≥82% Pb)

Lead(II) sulphide (≥82% Pb)

Glass bottle for solids, 2.500 g
Catalog # A10268.0E
Health
Irritant
Environment
Lead(II) sulphide (≥82% Pb)
Catalog # A10268.0E
CAS Number:  1314-87-0
Lead(II) sulphide (≥82% Pb)
Lead(II) sulphide (≥82% Pb)

Glass bottle for solids, 2.500 g

CAS Number:  1314-87-0
Price
193,00 €
Log in for availability
1 * 2.500 g
Product Details & Documents

CAS No.: 1314-87-0 Molecular Formula: PbS Formula Weight: 239.25 Refractive Index: 3.921 Odour: Odourless Melting Point: 1114° UN No.: 3077 MDL No.: MFCD00016280

Synonyms:  Lead sulphide
  • CAS Number:1314-87-0
  • Formula:̍PbS
  • MDL Number:MFCD00016280
  • Molecular Weight:239.27 g/mol
  • Merck Index:14,5421
  • Boiling Pt:1344 °C (1013 hPa)
  • Melting Pt:1114 °C
  • Density:7.5
  • Storage Temperature:Ambient
  • EINECS:215-246-6
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2.500 g

Specifications