VWR®, Polyester Films with Colour-Coded Marking Areas
Films with purple dot, Transparent, 77,5 µm, -
Catalog # 391-1267
Supplier: VWR Collection
Jump to:
Product Details & Documents
Identical to polyester films above, but include a colour dot and white writing area on each end tab for plate identification. End tabs with backing removed adhere to the ends of the plate and are perforated so they can be left in place even if the centre portion of the film is removed. Alternatively, one inscribed colour-coded end tab can be removed and pasted in a laboratory notebook while the other remains on the plate.
- Choice of five colours for easy plate identification
- Applications: Short-term storage, incubation and ELISA
- Recommended temperature range −40...+120 °C
Documents
Avantor Cell Culture Solutions.pdf VWR Sealing Film
Product Literature
Specifications
- Optical property:Transparent
- Sterile:-
- Description:Films with purple dot
- Thickness:77,5 µm



