Orders LinkContactUsLinkComponent
Warning signs
Harmful/irritating substance, PE, self-adhesive, 200×200 mm
Catalog # BRDY250265
Supplier:   Brady
Warning signs
Catalog # BRDY250265
Supplier:  Brady
Warning signs
Warning signs
Harmful/irritating substance, PE, self-adhesive, 200×200 mm
Price
4,10 €
Log in for availability
1 * 1 items
Product Details & Documents
These safety signs are for the purposes of accident prevention, fire protection, health hazard information and emergency evacuation.
  • Recommended by ISO 7010
  • Colour: Yellow/Black
  • Shape: Triangle
Specifications
  • Description:
    Harmful/irritating substance
  • Dimensions:
    200×200 mm
  • Material:
    PE, self-adhesive
  • Seq:
    23