pH buffers, ready to use
Technical buffer TPL-7/25, 7,00, Plastic bottle, 25
Catalog # WTWA108812
Supplier: WTW
Jump to:
Product Details & Documents
Every bottle has its own lot reference with the expiration date.
- Shelf life up to 3 years (unopened)
- Reference temperature at 25 °C
- Economic consumption
- Easiest handling by use of the dosing vessel
- One litre container with a separate compartment for the sample
Accuracy is ±0,03 pH
Certifications: Traceable to NIST.
Documents
Certificates
Search by Lot NumberCoC_WTWA108698_01.26-1 CoC_WTWA108698_05.23-1 CoC_WTWA108700_01-27.4 CoC_WTWA108700_01.24-2 CoC_WTWA108700_01.25-2
More
Sensors for Field and Laboratory.pdf
Additional Product Details
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:25
- Description:Technical buffer TPL-7/25
- pH value:7,00



