Orders LinkContactUsLinkComponent
Cover spectacles, 2800
Anti-UV (2 - 1,2), anti-scratch, anti-fog, Blue, Yellow PC
  MMMA2802
 :   3M
Cover spectacles, 2800
  MMMA2802
 :  3M
Cover spectacles, 2800
Cover spectacles, 2800
Anti-UV (2 - 1,2), anti-scratch, anti-fog, Blue, Yellow PC
12,00 €
1 * 1 items
These lightweight cover glasses (OTG) are designed to fit over most prescription spectacles with minimal interference. Available in three lens options. The green lens offers protection against infrared radiation arising from certain gas welding, gas cutting and brazing applications (IR shade 5).
  • Strong polycarbonate lens with anti-scratch coating
  • Adjustable temple length (four positions)
  • Inclinable lens for easy adjustment and maximum comfort
  • Soft low profile temples for comfort and minimum interaction with prescription spectacles
Weight: 38 g
 : EN 166, EN 170, EN 171 (for IR), FT 1
  • Type:
    Anti-UV (2 - 1,2), anti-scratch, anti-fog
  • Colour:
    Blue
  • Lenses:
    Yellow PC