Orders LinkContactUsLinkComponent
Cover spectacles, 2800
Cover spectacles, 2800
Anti-UV (2C - 1,2), anti-scratch, anti-fog, Blue, Clear PC
Catalog # 111-1603
Supplier:   3M
Cover spectacles, 2800
Catalog # 111-1603
Supplier:  3M
Cover spectacles, 2800
Cover spectacles, 2800
Anti-UV (2C - 1,2), anti-scratch, anti-fog, Blue, Clear PC
Price
12,00 €
Log in for availability
1 * 1 items
Product Details & Documents
These lightweight cover glasses (OTG) are designed to fit over most prescription spectacles with minimal interference. Available in three lens options. The green lens offers protection against infrared radiation arising from certain gas welding, gas cutting and brazing applications (IR shade 5).
  • Strong polycarbonate lens with anti-scratch coating
  • Adjustable temple length (four positions)
  • Inclinable lens for easy adjustment and maximum comfort
  • Soft low profile temples for comfort and minimum interaction with prescription spectacles
Weight: 38 g
Certifications: EN 166, EN 170, EN 171 (for IR), FT 1
Specifications
  • Type:
    Anti-UV (2C - 1,2), anti-scratch, anti-fog
  • Colour:
    Blue
  • Lenses:
    Clear PC