Order Entry
Export
Orders LinkContactUsLinkComponent
 

VWR®, Halogen Bulbs

Halogen bulb, 15 V, 150 W
Catalog # 631-1769
VWR®, Halogen Bulbs
Catalog # 631-1769
Supplier:  
VWR®, Halogen Bulbs
VWR®, Halogen Bulbs

Halogen bulb, 15 V, 150 W

Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Product Details & Documents
Specifications
  • Description:
    Halogen bulb
  • Seq:
    05
  • Power:
    150 W
  • Voltage:
    15 V