Orders LinkContactUsLinkComponent
Microplates, 96-well
Microplates, 96-well
Microplate, 96-well, F-bottom, without lid, Clear, 96
Catalog # BOET05.031.0100
Supplier:   BOETTGER PAUL
Microplates, 96-well
Catalog # BOET05.031.0100
Supplier:  BOETTGER PAUL
Microplates, 96-well
Microplates, 96-well
Microplate, 96-well, F-bottom, without lid, Clear, 96
Price
136,00 €
Log in for availability
1 * 100 items
Product Details & Documents
PS, optically clear, without lids. Microplates with a choice of different well types for different applications. The plates are fully compatible with standard reading and washing equipment.
  • Safe stacking, no danger of slipping
  • Gripping surfaces at the side of the plate
  • Frosted surface prevents errors from extraneous light sources
  • Alphanumerical well identification for easy sample location
  • Marking area on the side
  • Lids (optional) have condensation rings to reduce evaporation
  • Lid orientation corners reduce risk of cross-contamination
Specifications
  • No. of wells:
    96
  • Colour:
    Clear
  • Description:
    Microplate, 96-well, F-bottom, without lid
  • Seq:
    03