Order Entry
Export
Orders LinkContactUsLinkComponent
Dropping bottles, APONORM®

Dropping bottles, APONORM®

20 ml, 69 mm, 27,5 mm
Catalog # MUEL14.015.11
Supplier:   MUELLER KREMPEL
Dropping bottles, APONORM®
Catalog # MUEL14.015.11
Supplier:  MUELLER KREMPEL
Dropping bottles, APONORM®
Dropping bottles, APONORM®

20 ml, 69 mm, 27,5 mm

Price
21,50 €
Log in for availability
1 * 20 items
Product Details & Documents

Dropper bottles made of amber glass.

  • Good chemical and physical properties
  • For use in applications that do not involve high thermal stresses
  • Amber coloured to protect light sensitive substances

Thread according to DIN-18 and with dust protection plug, germ-free and hygienic shrink wrap packaging.

Specifications
  • Ext.Ø:
    27,5 mm
  • Capacity:
    20 ml
  • Height:
    69 mm