Order Entry
Export
Orders LinkContactUsLinkComponent
Slide wallets, FiloSlide™

Slide wallets, FiloSlide™

Slide wallets with four compartments, Transparent, 260×380 mm
Catalog # 720-2344
Supplier:   CELLPATH
Slide wallets, FiloSlide™
Catalog # 720-2344
Supplier:  CELLPATH
Slide wallets, FiloSlide™
Slide wallets, FiloSlide™

Slide wallets with four compartments, Transparent, 260×380 mm

Price
206,00 €
Log in for availability
1 * 100 items
Product Details & Documents

These slide folders feature 21 compartments for 76×26, 50×76 and 76×76 mm slides.

  • Hanging system
  • Storage in conventional filing cabinets
Specifications
  • W×D:
    260×380 mm
  • Colour:
    Transparent
  • Description:
    Slide wallets with four compartments