Orders LinkContactUsLinkComponent
Racks for pipette boxes
Racks for pipette boxes
Pipette rack, 6 pipette boxes, Stainless steel, 200×260×310 mm
Catalog # GLWAP06
Supplier:   GLW
Racks for pipette boxes
Catalog # GLWAP06
Supplier:  GLW
Racks for pipette boxes
Racks for pipette boxes
Pipette rack, 6 pipette boxes, Stainless steel, 200×260×310 mm
Price
141,12 €
Log in for availability
1 * 1 items
Product Details & Documents
These stainless steel racks are suitable for the storage of four or six GLW pipette boxes (A30, A40, A45 / S30, S40, S45).
Ordering information: Pipette boxes are not included.
Specifications
  • For:
    6 pipette boxes
  • Description:
    Pipette rack
  • Material:
    Stainless steel
  • W×D×H:
    200×260×310 mm