Order Entry
Export
Orders LinkContactUsLinkComponent
Hydrocortisone ≥98%

Hydrocortisone ≥98%

Glass bottle for solids, 25 g
Catalog # A16292.14
Health
Hydrocortisone ≥98%
Catalog # A16292.14
CAS Number:  50-23-7
Hydrocortisone ≥98%
Hydrocortisone ≥98%

Glass bottle for solids, 25 g

CAS Number:  50-23-7
Price
539,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 50-23-7 Molecular Formula: C21H30O5 Formula Weight: 362.47 Physical Form: Powder Appearance: White Odour: Odourless Melting Point: 213-215° Beilstein Reference: 1354819 Merck Reference: 14,4787 MDL No.: MFCD00011654

A natural corticosteroid and anti-inflammatory compound

  • CAS Number:50-23-7
  • Formula:C₂₁H₃₀O₅
  • MDL Number:MFCD00011654
  • Molecular Weight:362.47 g/mol
  • Merck Index:14,4787
  • Boiling Pt:567 °C (1013 hPa)
  • Melting Pt:213…215 °C
  • Density:1.4 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-020-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications