Order Entry
Export
Orders LinkContactUsLinkComponent
Dextrin, precipitated by alcohol

Dextrin, precipitated by alcohol

Glass bottle for solids, 500 g
Catalog # A15717.36
Dextrin, precipitated by alcohol
Catalog # A15717.36
CAS Number:  9004-53-9
Dextrin, precipitated by alcohol
Dextrin, precipitated by alcohol

Glass bottle for solids, 500 g

CAS Number:  9004-53-9
Price
105,00 €
Log in for availability
1 * 500 g
Product Details & Documents

CAS No.: 9004-53-9 Merck Reference: 14,2953 MDL No.: MFCD00081554

Synonyms:  Dextrin
  • CAS Number:9004-53-9
  • Formula:(C₆H₁₂O₆)n
  • MDL Number:MFCD00081554
  • Molecular Weight:180.16 g/mol
  • Merck Index:14,2953
  • Density:1.45 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:232-675-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications