Orders LinkContactUsLinkComponent
Universal replacement parts, Omnifit®
Universal replacement parts, Omnifit®
Replacement frits, PE, for Omnifit® columns, 25 µm porosity, 15 mm bore
Catalog # OMNI006FR-15-25-PE
Supplier:   OMNIFIT
Universal replacement parts, Omnifit®
Catalog # OMNI006FR-15-25-PE
Supplier:  OMNIFIT
Universal replacement parts, Omnifit®
Universal replacement parts, Omnifit®
Replacement frits, PE, for Omnifit® columns, 25 µm porosity, 15 mm bore
Price
236,00 €
Log in for availability
1 * 20 items
Product Details & Documents
Universal replacement parts for all Omnifit® labware columns which gives complete flexibility to replace lost or worn parts. Compatible with BenchMark, SolventPlus and HiT column ranges.
Specifications
  • Description:
    Replacement frits, PE, for Omnifit® columns, 25 µm porosity, 15 mm bore
  • Seq:
    44