Universal replacement parts, Omnifit®
Replacement frits, PE, for Omnifit® columns, 25 µm porosity, 6,6 mm bore
OMNI006FR-06-25-PE
: OMNIFIT
Universal replacement parts for all Omnifit® labware columns which gives complete flexibility to replace lost or worn parts. Compatible with BenchMark, SolventPlus and HiT column ranges.
- Description:Replacement frits, PE, for Omnifit® columns, 25 µm porosity, 6,6 mm bore
- Seq:41