Order Entry
Export
Orders LinkContactUsLinkComponent
 

Transfer pipettes, Samco™ Extra Long

24,1 ml, 20, 10,7 ml, 8,6 mm, 300 mm, +, Individually wrapped
Catalog # SAMC263-1S
   Low Manufacturing Impact
Supplier:   SAMCO
Transfer pipettes, Samco™ Extra Long
Catalog # SAMC263-1S
Supplier:  SAMCO
   Low Manufacturing Impact
Transfer pipettes, Samco™ Extra Long
Transfer pipettes, Samco™ Extra Long

24,1 ml, 20, 10,7 ml, 8,6 mm, 300 mm, +, Individually wrapped

Price
59,20 €
Log in for availability
1 * 100 items
Product Details & Documents

These extra long PE pipettes are ideal for safe and easy sampling from large containers.

  • Excellent for tissue culture applications
  • General purpose pipettes are ideal for routine sampling of serum and plasma
  • Well suited for blood culture and urine analysis

Perfect for tissue culture applications, these pipettes are designed with a built-in pipette bulb that eliminates the need for plugging, packaging, autoclaving and prevents cross-contamination.

Specifications
  • Ext.Ø:
    8,6 mm
  • Packed:
    Individually wrapped
  • Sterile:
    +
  • Capacity:
    24,1 ml
  • Length:
    300 mm
  • Seq:
    05
  • Drops per ml:
    20
  • Bulb draw:
    10,7 ml
  • Cat. No.:
    SAMC263-1S
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact