Transfer pipettes, Samco™ Extra Long
24,1 ml, 20, 10,7 ml, 8,6 mm, 300 mm, -, 100 per pack
Catalog # SAMC263
Supplier: SAMCO
Jump to:
Product Details & Documents
These extra long PE pipettes are ideal for safe and easy sampling from large containers.
- Excellent for tissue culture applications
- General purpose pipettes are ideal for routine sampling of serum and plasma
- Well suited for blood culture and urine analysis
Perfect for tissue culture applications, these pipettes are designed with a built-in pipette bulb that eliminates the need for plugging, packaging, autoclaving and prevents cross-contamination.
Specifications
- Ext.Ø:8,6 mm
- Packed:100 per pack
- Sterile:-
- Capacity:24,1 ml
- Length:300 mm
- Seq:04
- Drops per ml:20
- Bulb draw:10,7 ml
- Cat. No.:SAMC263
- Environmental certification:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Low Manufacturing Impact



