Order Entry
Export
Orders LinkContactUsLinkComponent
Transfer pipettes, Samco™ Extra Long

Transfer pipettes, Samco™ Extra Long

5,6 ml, 25, 2,3 ml, 5,0 mm, 225 mm, +, 20 per pack
Catalog # SAMC262-20S
   Low Manufacturing Impact
Supplier:   SAMCO
Transfer pipettes, Samco™ Extra Long
Catalog # SAMC262-20S
Supplier:  SAMCO
   Low Manufacturing Impact
Transfer pipettes, Samco™ Extra Long
Transfer pipettes, Samco™ Extra Long

5,6 ml, 25, 2,3 ml, 5,0 mm, 225 mm, +, 20 per pack

Price
45,30 €
Log in for availability
1 * 400 items
Product Details & Documents

These extra long PE pipettes are ideal for safe and easy sampling from large containers.

  • Excellent for tissue culture applications
  • General purpose pipettes are ideal for routine sampling of serum and plasma
  • Well suited for blood culture and urine analysis

Perfect for tissue culture applications, these pipettes are designed with a built-in pipette bulb that eliminates the need for plugging, packaging, autoclaving and prevents cross-contamination.

Specifications
  • Ext.Ø:
    5,0 mm
  • Packed:
    20 per pack
  • Sterile:
    +
  • Capacity:
    5,6 ml
  • Length:
    225 mm
  • Seq:
    03
  • Drops per ml:
    25
  • Bulb draw:
    2,3 ml
  • Cat. No.:
    SAMC262-20S
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact