Order Entry
Export
Orders LinkContactUsLinkComponent
 

3,3'-Thiodipropionic acid ≥98%

Glass bottle for solids, 250 g
Catalog # A17220.30
Irritant
3,3'-Thiodipropionic acid ≥98%
Catalog # A17220.30
CAS Number:  111-17-1
3,3'-Thiodipropionic acid ≥98%
3,3'-Thiodipropionic acid ≥98%

Glass bottle for solids, 250 g

CAS Number:  111-17-1
Price
34,30 €
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 98% CAS No.: 111-17-1 Molecular Formula: C6H10O4S Formula Weight: 178.21 Melting Point: 127-134° Beilstein Reference: 1210299 Merck Reference: 14,9332 MDL No.: MFCD00002781

Synonyms:  3,3-Thiodipropionic acid
  • CAS Number:111-17-1
  • Formula:C₆H₁₀O₄S
  • MDL Number:MFCD00002781
  • Molecular Weight:178.21 g/mol
  • Merck Index:14,9332
  • Boiling Pt:347 °C (1013 hPa)
  • Melting Pt:127…134 °C
  • Density:1.496 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:203-841-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications