Order Entry
Export
Orders LinkContactUsLinkComponent
Laurinaldehyde 95% stabilised

Laurinaldehyde 95% stabilised

Glass bottle for solids, 100 g
Catalog # A15611.22
Irritant
Laurinaldehyde 95% stabilised
Catalog # A15611.22
CAS Number:  112-54-9
Laurinaldehyde 95% stabilised
Laurinaldehyde 95% stabilised

Glass bottle for solids, 100 g

CAS Number:  112-54-9
Price
66,50 €
Log in for availability
1 * 100 g
Product Details & Documents

Stabilized

Purity: 95% CAS No.: 112-54-9 Molecular Formula: CH3(CH2)10CHO Formula Weight: 184.32 Flash Point: 101°(214°F) Refractive Index: 1.435 Melting Point: 12° Boiling Point: 184-186°/100mm Beilstein Reference: 1703917 MDL No.: MFCD00007017

Synonyms:  Lauric aldehyde, Dodecylaldehyde, Dodecanal
  • CAS Number:112-54-9
  • Formula:C₁₂H₂₄O
  • MDL Number:MFCD00007017
  • Molecular Weight:184.32 g/mol
  • Boiling Pt:184…186 °C (100 mmHg)
  • Melting Pt:12 °C
  • Density:0.835
  • Flash Pt:101 °C (214 °F)
  • Storage Temperature:Ambient
  • EINECS:203-983-6
  • ADR:9,III
  •  :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
  • UN:3082
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications