Orders LinkContactUsLinkComponent
Lead(II) sulphide (≥82% Pb)
Glass bottle for solids, 500 g
  A10268.36
Lead(II) sulphide (≥82% Pb)
  A10268.36
 :  1314-87-0
Lead(II) sulphide (≥82% Pb)
Lead(II) sulphide (≥82% Pb)
Glass bottle for solids, 500 g
 :  1314-87-0
55,00 €
1 * 500 g
CAS No.: 1314-87-0 Molecular Formula: PbS Formula Weight: 239.25 Refractive Index: 3.921 Odour: Odourless Melting Point: 1114° UN No.: 3077 MDL No.: MFCD00016280
 :  Lead sulphide
  •  :1314-87-0
  •  :̍PbS
  •  :MFCD00016280
  •  :239.27 g/mol
  •  :14,5421
  •  :1344 °C (1013 hPa)
  •  :1114 °C
  •  :7.5
  •  :Ambient
  •  :215-246-6
  •  :3077
  •  :9,III
  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g