Orders LinkContactUsLinkComponent
SYPRO® Ruby Protein Gel Stain
SYPRO® Ruby Protein Gel Stain
SYPRO® Ruby Protein Gel Stain, 200 mL
Catalog # 730-1307
Supplier:   LONZA
SYPRO® Ruby Protein Gel Stain
Catalog # 730-1307
Supplier:  LONZA
CAS Number:  
SYPRO® Ruby Protein Gel Stain
SYPRO® Ruby Protein Gel Stain, 200 mL
CAS Number:  
Price
194,00 €
Log in for availability
1 * 200 mL
Product Details & Documents
SYPRO™ Ruby Protein Gel Stain is highly sensitive, simple to use fluorescent stain that can accurately quantitate protein expression levels, and is compatible with standard fluorescent visualization systems and downstream identification techniques such as mass spectrometry.
  • Visualize proteins prior to transfer - Does not interfere with protein activity
  • Versatile - Excellent for staining native proteins
  • Safe - No acids or organic solvents necessary
  • Sensitive - Detects as little as 4-8 ng protein per band
Specifications
Specifications
  • Pk:
    200 mL
  • Description:
    SYPRO® Ruby Protein Gel Stain
Specifications