DNA ladder
100 bp ladder, 10 fragments, with 100 ng/µl concentration, 100 Assays
LONZ50321
: LONZA
Thse DNA ladders provides rapid and easy size determination of the DNA samples. Each band has an ideal 50 % G-C content for precise sizing and sharp resolution. It is free from contaminating plasmid DNA with minimal background.
- Pk:100 Assays
- Description:100 bp ladder, 10 fragments, with 100 ng/µl concentration