Order Entry
Export
Orders LinkContactUsLinkComponent
Storage plates, 384-well, MASTERBLOCK®

Storage plates, 384-well, MASTERBLOCK®

Deep well, V-bottom, sterile, Natural
Catalog # 738-0501
Supplier:   Greiner Bio-One
Storage plates, 384-well, MASTERBLOCK®
Catalog # 738-0501
Supplier:  Greiner Bio-One
Storage plates, 384-well, MASTERBLOCK®
Storage plates, 384-well, MASTERBLOCK®

Deep well, V-bottom, sterile, Natural

Price
746,00 €
Log in for availability
1 * 60 items
Product Details & Documents

PP, without lid. Ideal for compound libraries and sample storage.

  • Biologically inert, resistant to commonly used solvents (such as DMSO, acetonitrile or acetone) and temperature resistant (−196 °C to +121 °C)
  • Uniform external dimensions and tight tolerances make robotic handling easier
  • Can be easily sealed with adhesive films, heat seals or CapMats
  • Alphanumeric well coding
  • Conical well shape enables precise pipetting and rounded square well design combines flexible working volume (20 to 225 µl) with reduced wicking and bubbling

Colour: Natural

Colour: Natural

Specifications
  • Colour:
    Natural
  • Description:
    Deep well, V-bottom, sterile