Order Entry
Export
Orders LinkContactUsLinkComponent
Ethanol absolute ≥99.5%, TechniSolv®, pure

Ethanol absolute ≥99.5%, TechniSolv®, pure

Metal drum, 200 L
Catalog # 83813.550
Flammable
Irritant
   Safer Human and Environmental Health
Supplier:   VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Catalog # 83813.550
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  64-17-5
Ethanol absolute ≥99.5%, TechniSolv®, pure
Ethanol absolute ≥99.5%, TechniSolv®, pure

Metal drum, 200 L

CAS Number:  64-17-5
Price
1 570,00 €
Log in for availability
1 * 200 L
Product Details & Documents

Caution: WARNING : This alcohol is regulated. Please contact VWR. For duty paid orders in the UK please add DP to the product code.

Synonyms:  Ethyl alcohol
  • CAS Number:64-17-5
  • Formula:C₂H₅OH
  • MDL Number:MFCD00003568
  • Molecular Weight:46.07 g/mol
  • Merck Index:12,03806
  • Boiling Pt:78.3 °C (1013 hPa)
  • Melting Pt:–117 °C
  • Density:0.7895 g/cm³ (20 °C)
  • Flash Pt:12 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-578-6
  • UN:1170
  • ADR:3,II
  • REACH:01-2119457610-43
Specifications

Specification Test Results

  • Assay (V/V):
    Min. 99.5 %
  • Density (20/20):
    0.788 - 0.794
  • Water:
    Max. 0.5 %

Specifications

  • Pack type:
    Metal drum
  • Pk:
    200 L
  • Cat. No.:
    83813.550
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications