Ethanol ≥96% (v/v), TechniSolv®, pure
Metal drum, 200 L
Supplier: VWR Chemicals
Ethanol ≥96% (v/v), TechniSolv®, pure
Catalog # 83804.550
Supplier: VWR Chemicals
CAS Number: 64-17-5
Jump to:
Product Details & Documents
Caution: WARNING : This alcohol is regulated. Please contact VWR. For duty paid orders in the UK please add DP to the product code.
Synonyms: Ethyl alcohol
- CAS Number:64-17-5
- Formula:C₂H₅OH
- MDL Number:MFCD00003568
- Molecular Weight:46.07 g/mol
- Boiling Pt:78.3 °C (1013 hPa)
- Melting Pt:–117 °C
- Density:0.7895 g/cm³ (20 °C)
- Flash Pt:12 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-578-6
- UN:1170
- ADR:3,II
- REACH:01-2119457610-43
Documents
Certificates
Search by Lot Number10L Plastic drum for liquids DIN 51 UN (ZPACK13951000) 225L Metal drum UN (ZPACE8010050) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic jerrycan for liquids DIN 42 (ZPACK13951025)
Packaging Information
P20_VWRC83804.360_SPEC_EN.pdf P20_VWRC83804.410_SPEC_EN.pdf P20_VWRC83804.440_SPEC_EN.pdf P20_VWRC83804.550_SPEC_EN.pdf spec_VWRC83804.360
Product specification
More
VWR Chemicals Big Packs and Bulks VWR Denatured Ethanol
Product Literature
Specifications
Specification Test Results
- Assay:Min. 95 %
- Assay (V/V):Min. 95 %
- Density (20/20):0.805 - 0.811
Specifications
- Pack type:Metal drum
- Pk:200 L
- Cat. No.:83804.550
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
- Environmentally Preferable:Safer Human and Environmental Health



