Ethanol ≥96% (v/v), TechniSolv®, pure
Plastic jerrican, 5 L
Caution: WARNING : This alcohol is regulated. Please contact VWR. For duty paid orders in the UK please add DP to the product code.
: Ethyl alcohol
- :64-17-5
- :C₂H₅OH
- :MFCD00003568
- :46.07 g/mol
- :78.3 °C (1013 hPa)
- :–117 °C
- :0.7895 g/cm³ (20 °C)
- :12 °C (closed cup)
- :Ambient
- :200-578-6
- :1170
- :3,II
- :01-2119457610-43
Documents
10L Plastic drum for liquids DIN 51 UN (ZPACK13951000) 225L Metal drum UN (ZPACE8010050) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic jerrycan for liquids DIN 42 (ZPACK13951025)
Packaging Information
P20_VWRC83804.360_SPEC_EN.pdf P20_VWRC83804.410_SPEC_EN.pdf P20_VWRC83804.440_SPEC_EN.pdf P20_VWRC83804.550_SPEC_EN.pdf spec_VWRC83804.360
Product specification
VWR Chemicals Big Packs and Bulks VWR Denatured Ethanol
Product Literature
- Assay:Min. 95 %
- Assay (V/V):Min. 95 %
- Density (20/20):0.805 - 0.811
- Pack type:Plastic jerrican
- Pk:5 L



