Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Color Comparison Tubes (Case of 6) 1 * 1 items
KIML45290-154
:
KIMBLEKONTES
Color Comparison Tubes (Case of 6) 1 * 1 items
KIML45290-154
:
KIMBLEKONTES