Order Entry
Export
Orders LinkContactUsLinkComponent
D-(+)-Sucrose, Millipore®

D-(+)-Sucrose, Millipore®

Glass bottle for solids, 1 kg
Catalog # SIAL84100-1KG
Supplier:   Merck
D-(+)-Sucrose, Millipore®
Catalog # SIAL84100-1KG
Supplier:  Merck
CAS Number:  57-50-1
D-(+)-Sucrose, Millipore®
D-(+)-Sucrose, Millipore®

Glass bottle for solids, 1 kg

CAS Number:  57-50-1
Price
70,80 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose
  • CAS Number:57-50-1
  • Formula:C₁₂H₂₂O₁₁
  • MDL Number:MFCD00006626
  • Molecular Weight:342.3 g/mol
  • Merck Index:13,08966
  • Melting Pt:169…170 °C
  • Density:1.29 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:200-334-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications