Order Entry
Export
Orders LinkContactUsLinkComponent
α-Cyclodextrin, Sigma-Aldrich®

α-Cyclodextrin, Sigma-Aldrich®

Glass bottle for solids, 100 g
Catalog # SIAL28705-100G
Supplier:   Merck
α-Cyclodextrin, Sigma-Aldrich®
Catalog # SIAL28705-100G
Supplier:  Merck
CAS Number:  10016-20-3
α-Cyclodextrin, Sigma-Aldrich®

Glass bottle for solids, 100 g

CAS Number:  10016-20-3
Price
181,00 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:10016-20-3
  • Formula:C₃₆H₆₀O₃₀
  • MDL Number:MFCD00078207
  • Molecular Weight:972.84 g/mol
  •  :M-Clarity™ quality level = MQ200
  • EINECS:233-007-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications