Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
GIEMSA STAIN, MODIFIED SOLUTION 1 * 1 L
GIEMSA STAIN, MODIFIED SOLUTION 1 * 1 L
Print
GIEMSA STAIN, MODIFIED SOLUTION 1 * 1 L
Catalog # SIAL48900-1L-F
Supplier:
Merck
GIEMSA STAIN, MODIFIED SOLUTION 1 * 1 L
Catalog # SIAL48900-1L-F
Supplier:
Merck
CAS Number:
Expand image
Return to Top
GIEMSA STAIN, MODIFIED SOLUTION 1 * 1 L
Add to cart
CAS Number:
Price
135,00 €
Log in
for availability
Unit of Measure
1 * 1 L
-
+
Add to cart
Return Policy
Product Details & Documents