Cycloheximide, Sigma-Aldrich®
Glass bottle for solids, 5 g
SIAL01810-5G
: Merck
: Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
- :66-81-9
- :C₁₅H₂₃NO₄
- :MFCD00082346
- :281.35 g/mol
- :Ambient
- :6.1,I
- :M-Clarity™ quality level = MQ200
- :200-636-0
- :2811
- Pack type:Glass bottle for solids
- Pk:5 g



