Diffusion tubes, with direct indication, for TWA measurements
Butadiene 10/a-D, 10 - 300 ppm / 1 hr
Catalog # DRAE8101161
Supplier: Dräger
Diffusion tubes, with direct indication, for TWA measurements
Catalog # DRAE8101161
Supplier: Dräger
Jump to:
Product Details & Documents
Long-term measurements with diffusion tubes provide integrated measurements that represent the average concentration during the sampling period.
- Measurements performed between one and eight H
- Tubes can be used as personal, or area monitors
- Determine the weight average concentration
Specifications
- Range:10 - 300 ppm / 1 hr
- Description:Butadiene 10/a-D
- Seq:04
Accessories
Product Information
- Catalog #AvailabilityPrice
- Holders