Orders LinkContactUsLinkComponent
Microplates, 1536-well
Microplates, 1536-well
Solid bottom, clear, Clear, 1536, -
Catalog # 738-0080
   Low Manufacturing Impact
Supplier:   Greiner Bio-One
Microplates, 1536-well
Catalog # 738-0080
Supplier:  Greiner Bio-One
   Low Manufacturing Impact
Microplates, 1536-well
Microplates, 1536-well
Solid bottom, clear, Clear, 1536, -
Price
2 300,00 €
Log in for availability
1 * 60 items
Product Details & Documents
PS, without lid. Ideal for high throughput screening applications, these plates are available as clear bottom variants (in clear PS) and black or white in LUMITRAC™ or FLUOTRAC™ quality.
  • HiBase versions designed for top reading instruments, µClear® LoBase plates for bottom reading instruments
  • Solid bottom plates available with high binding (MICROLON™ 600, LUMITRAC™ 600 or FLUOTRAC™ 600) or medium binding (LUMITRAC™ 200 or FLUOTRAC™ 200) surfaces
  • Rounded square well design combines the advantages of the square well (flexible working volume, 3 to 10 µl) with the advantages of a round well (reduced wicking and bubbling)
  • Free of DNase, RNase and human DNA, and are non-pyrogenic
Working volume: 3 - 10 µl
Specifications
  • No. of wells:
    1536
  • Sterile:
    -
  • Colour:
    Clear
  • Description:
    Solid bottom, clear
  • Cat. No.:
    738-0080
  • Environmental certification:
    ISO 14001 (241002); ISO 50001 (241002)
  • Environmentally Preferable:
    Low Manufacturing Impact