VWR® Flasks, Flat Bottom, with Standard Ground Joint
100 ml, 103 mm, 64 mm, 29/32 NS
Borosilicate 3.3 glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Inexpensive alternative for a wide range of laboratory applications. With white labelling area and imprinted catalogue number for convenient reordering.
DIN 12348
: DIN 12348
- Ext.Ø:64 mm
- Socket size:29/32 NS
- Capacity:100 ml
- Height:103 mm
- Seq:04



