Order Entry
Export
Orders LinkContactUsLinkComponent
Reservoir system

Reservoir system

Reservoir, quarter, 40 ml, +
Catalog # BECL372790
Supplier:   Beckman Coulter
Reservoir system
Catalog # BECL372790
Supplier:  Beckman Coulter
Reservoir system
Reservoir system

Reservoir, quarter, 40 ml, +

Price
247,00 €
Log in for availability
1 * 48 items
Product Details & Documents

This modular reservoir system holds liquids for pipetting with a manual pipettor or liquid handling systems including the full suite of Biomek liquid handlers.

  • Half and quarter reservoirs must be used with a frame
  • Full reservoirs hold up to 150 ml of liquid, and do not require a frame
  • Each frame can accommodate up to four quarter reservoirs or two half reservoirs
  • All reservoirs are sterile and made of PP

Not for use with 96 and 384 multi channel pipetting heads.

Documents

Specifications
  • Sterile:
    +
  • Capacity:
    40 ml
  • Description:
    Reservoir, quarter
  • Seq:
    1