Orders LinkContactUsLinkComponent
Volumetric flasks, class A, white markings
Volumetric flasks, class A, white markings
300 ml, 14/23, 0,150 ± ml
Catalog # WITG3.670.030
Supplier:   witeg Labortechnik
Volumetric flasks, class A, white markings
Catalog # WITG3.670.030
Supplier:  witeg Labortechnik
Volumetric flasks, class A, white markings
Volumetric flasks, class A, white markings
300 ml, 14/23, 0,150 ± ml
Price
40,90 €
Log in for availability
1 * 1 items
Product Details & Documents
Borosilicate glass 3.3, amber, with glass stopper.
  • White markings
  • Conformity certified (DE-M marking)
  • Inclusive batch certificate
Certifications: DIN EN ISO 1042
Specifications
  • NS:
    14/23
  • Capacity:
    300 ml
  • Seq:
    09
  • Tolerance:
    0,150 ± ml