Order Entry
Export
Orders LinkContactUsLinkComponent
Chemists spatulas

Chemists spatulas

Flexible, elastic chemists spatula, 220 mm, 130 mm
Catalog # HAMMHSN202-13
Supplier:   HAMMACHER KARL
Chemists spatulas
Catalog # HAMMHSN202-13
Supplier:  HAMMACHER KARL
Chemists spatulas
Chemists spatulas

Flexible, elastic chemists spatula, 220 mm, 130 mm

Price
77,10 €
Log in for availability
1 * 1 items
Product Details & Documents

Flexible, elastic chemists spatulas with stainless steel blade.

Specifications
  • Description:
    Flexible, elastic chemists spatula
  • Length:
    220 mm
  • Blade length:
    130 mm