Cary source lamps
Lamp, deuterium, UV, for Cary 4000/5000/6000i
Catalog # VARI110713990
Supplier: VARIAN
Jump to:
Product Details & Documents
Source lamps for Cary spectrophotometers are fully modular and interchangeable. A range of UV, xenon and visible lamps are available.
- Fully modular and compatible with Cary spectrophotometers
The UV, xenon and visible lamps are fully modular, allowing you to select and change between light sources according to your experimental requirements.
Specifications
- Description:Lamp, deuterium, UV, for Cary 4000/5000/6000i
- Seq:01