Order Entry
Export
Orders LinkContactUsLinkComponent
 

Triclosan, Reference Standards

Triclosan Related Compounds Mixture A, USP reference standard, 1.2 mL/ampule; 3 ampules, Ampule, 3 Ampoul
Catalog # 1682217.
Supplier:   USP
Triclosan, Reference Standards
Catalog # 1682217.
Supplier:  USP
CAS Number:  
Triclosan, Reference Standards

Triclosan Related Compounds Mixture A, USP reference standard, 1.2 mL/ampule; 3 ampules, Ampule, 3 Ampoul

CAS Number:  
Price
1 190,00 €
Log in for availability
1 * 3 Ampoul
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

 

Specifications

Specifications

  • Pack type:
    Ampule
  • Pk:
    3 Ampoul
  • Standards:
    USP reference standard
  • Description:
    Triclosan Related Compounds Mixture A
  • Pk Info:
    1.2 mL/ampule; 3 ampules
  • UN No.:
    1230
  • Cat. No.:
    1682217.
  • Description:
    Triclosan Related Compounds Mixture A

Specifications